Assay Details
| Gene Symbol: Mpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010824.2 | NP_034954.2 | 2 - 3 | 373 | 67 | |
| GenBank mRNA | AK160229.1 | - | 1 - 2 | 94 | 67 | |
| AK220229.1 | - | 1 - 2 | 146 | 67 | ||
| AY500847.1 | - | 2 - 3 | 127 | 67 | ||
| BC053912.1 | - | 1 - 2 | 93 | 30044601 | 67 | |
| CA547566.1 | - | 2 - 3 | 327 | 67 | ||
| CN661559.1 | - | 1 - 2 | 55 | 67 | ||
| X15313.1 | - | 2 - 3 | 583 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpo
Genomic Map