Assay Details
| Gene Symbol: Mpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010824.2 | NP_034954.2 | 4 - 5 | 638 | 73 | |
| GenBank mRNA | AK160229.1 | - | 3 - 4 | 364 | 73 | |
| AK220229.1 | - | 3 - 4 | 411 | 73 | ||
| AY500847.1 | - | 4 - 5 | 392 | 73 | ||
| BC053912.1 | - | 3 - 4 | 358 | 30044601 | 73 | |
| CN661559.1 | - | 3 - 4 | 320 | 73 | ||
| X15313.1 | - | 4 - 5 | 848 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpo
Genomic Map