Assay Details
| Gene Symbol: Mpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010824.2 | NP_034954.2 | 12 - 13 | 2244 | 87 | |
| GenBank mRNA | AK160229.1 | - | 11 - 12 | 1972 | 87 | |
| AK220229.1 | - | 11 - 12 | 2017 | 87 | ||
| AY500847.1 | - | 12 - 13 | 1998 | 87 | ||
| BC053912.1 | - | 11 - 12 | 1964 | 30044601 | 87 | |
| X15313.1 | - | 12 - 13 | 2454 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpo
Genomic Map