Assay Details
| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011074.2 | NP_035204.2 | 5 - 6 | 671 | 62 | |
| GenBank mRNA | AF033655.1 | - | 5 - 6 | 630 | 62 | |
| AK045083.1 | - | 5 - 6 | 672 | 62 | ||
| AK051283.1 | - | 5 - 6 | 645 | 62 | ||
| AK051411.1 | - | 5 - 6 | 614 | 62 | ||
| AK083269.1 | - | 4 - 5 | 826 | 62 | ||
| AK083790.1 | - | 5 - 6 | 614 | 62 | ||
| AK087398.1 | - | 4 - 5 | 442 | 62 | ||
| AK129226.1 | - | 5 - 6 | 498 | 62 | ||
| AK138437.1 | - | 4 - 5 | 912 | 62 | ||
| AK139386.1 | - | 4 - 5 | 832 | 62 | ||
| AK141535.1 | - | 4 - 5 | 600 | 62 | ||
| AK165149.1 | - | 4 - 5 | 516 | 62 | ||
| BC068134.1 | - | 4 - 5 | 875 | 6834018 | 62 | |
| U62391.1 | - | 4 - 5 | 525 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk14
Genomic Map