Assay Details
| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011074.2 | NP_035204.2 | 9 - 10 | 1077 | 102 | |
| GenBank mRNA | AF033655.1 | - | 9 - 10 | 1036 | 102 | |
| AK051283.1 | - | 9 - 10 | 1051 | 102 | ||
| AK083269.1 | - | 8 - 9 | 1232 | 102 | ||
| AK087398.1 | - | 8 - 10 | 102 | |||
| AK129226.1 | - | 9 - 10 | 904 | 102 | ||
| AK138437.1 | - | 8 - 9 | 1318 | 102 | ||
| AK139386.1 | - | 8 - 9 | 1238 | 102 | ||
| AK141535.1 | - | 8 - 9 | 1006 | 102 | ||
| AK165149.1 | - | 8 - 9 | 922 | 102 | ||
| BC068134.1 | - | 8 - 9 | 1281 | 6834018 | 102 | |
| U62391.1 | - | 8 - 9 | 931 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk14
Genomic Map