Assay Details
| Gene Symbol: Phf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011078.3 | NP_035208.2 | 2 - 3 | 297 | 62 | |
| GenBank mRNA | AB221591.1 | - | 2 - 3 | 181 | 62 | |
| AF043726.1 | - | 2 - 3 | 297 | 62 | ||
| BC051633.1 | - | 2 - 3 | 295 | 6518591 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phf2
Genomic Map