Assay Details
| Gene Symbol: Phf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011078.3 | NP_035208.2 | 7 - 8 | 1073 | 95 | |
| GenBank mRNA | AB221591.1 | - | 7 - 8 | 957 | 95 | |
| AF043726.1 | - | 7 - 8 | 1073 | 95 | ||
| AK153906.1 | - | 7 - 8 | 1223 | 95 | ||
| AK172995.1 | - | 5 - 6 | 780 | 95 | ||
| BC051633.1 | - | 7 - 8 | 1071 | 6518591 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phf2
Genomic Map