Assay Details
| Gene Symbol: Phf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011078.3 | NP_035208.2 | 12 - 13 | 1805 | 63 | |
| GenBank mRNA | AB221590.1 | - | 1 - 2 | 191 | 63 | |
| AB221591.1 | - | 12 - 13 | 1689 | 63 | ||
| AF043726.1 | - | 12 - 13 | 1805 | 63 | ||
| AK172995.1 | - | 10 - 11 | 1512 | 63 | ||
| BC051633.1 | - | 12 - 13 | 1803 | 6518591 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phf2
Genomic Map