Assay Details
Gene Symbol: Sptlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011479.3 | NP_035609.1 | 2 - 3 | 377 | 113 | |
GenBank mRNA | AK044917.1 | - | 2 - 3 | 378 | 113 | |
AK057644.1 | - | 2 - 3 | 413 | 113 | ||
AK089019.1 | - | 2 - 3 | 371 | 113 | ||
AK159346.1 | - | 2 - 3 | 378 | 113 | ||
BC003227.1 | - | 2 - 3 | 410 | 3483158 | 113 | |
U27455.1 | - | 2 - 3 | 345 | 113 | ||
X95642.1 | - | 2 - 3 | 348 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
