Assay Details
| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011479.3 | NP_035609.1 | 3 - 4 | 531 | 121 | |
| GenBank mRNA | AK044917.1 | - | 3 - 4 | 532 | 121 | |
| AK057644.1 | - | 3 - 4 | 567 | 121 | ||
| AK089019.1 | - | 3 - 4 | 525 | 121 | ||
| AK159346.1 | - | 3 - 4 | 532 | 121 | ||
| BC003227.1 | - | 3 - 4 | 564 | 3483158 | 121 | |
| U27455.1 | - | 3 - 4 | 499 | 121 | ||
| X95642.1 | - | 3 - 4 | 502 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc2
Genomic Map