Assay Details
| Gene Symbol: Stat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037598.1 | - | 2 - 3 | 246 | 154 | |
| AK132675.1 | - | 4 - 5 | 433 | 154 | ||
| AK157083.1 | - | 3 - 4 | 307 | 154 | ||
| BC052907.1 | - | 3 - 4 | 312 | 154 | ||
| BC098499.1 | - | 3 - 4 | 300 | 30074608 | 154 | |
| BY750146.1 | - | 3 - 4 | 298 | 154 | ||
| U06923.1 | - | 3 - 4 | 303 | 154 | ||
| U09351.1 | - | 3 - 4 | 757 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stat4
Genomic Map