Assay Details
| Gene Symbol: Vipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011703.4 | NP_035833.2 | 5 - 6 | 638 | 67 | |
| GenBank mRNA | AF266282.1 | - | 5 - 6 | 520 | 67 | |
| AK052465.1 | - | 5 - 6 | 639 | 67 | ||
More Information
Set Membership: |
|

Vipr1
Genomic Map