Assay Details
| Gene Symbol: Vipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011703.4 | NP_035833.2 | 10 - 11 | 1149 | 75 | |
| GenBank mRNA | AF266282.1 | - | 10 - 11 | 1031 | 75 | |
| AK052465.1 | - | 10 - 11 | 1150 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vipr1
Genomic Map