Assay Details
| Gene Symbol: Mok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011973.2 | NP_036103.1 | 1 - 2 | 195 | 65 | |
| GenBank mRNA | AB022695.1 | - | 1 - 2 | 195 | 65 | |
| BC049604.1 | - | 1 - 2 | 183 | 65 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB022695.1,NM_011973.2 transcripts.
Set Membership: |
|

Mok
Genomic Map