Assay Details
Gene Symbol: Mok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011973.2 | NP_036103.1 | 3 - 4 | 392 | 117 | |
GenBank mRNA | AB022695.1 | - | 3 - 4 | 392 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
