Assay Details
| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013691.2 | NP_038719.2 | 17 | 2019 | 72 | |
| GenBank mRNA | BC053023.1 | - | 17 | 1973 | 6401682 | 72 |
| BC060104.1 | - | 15 | 3011 | 72 | ||
| L04302.1 | - | 17 | 1999 | 72 | ||
| L24434.1 | - | 17 | 1996 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Thbs3
Genomic Map