Assay Details
| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013691.2 | NP_038719.2 | 21 - 22 | 2734 | 86 | |
| GenBank mRNA | BC053023.1 | - | 21 - 22 | 2688 | 6401682 | 86 |
| L04302.1 | - | 21 - 22 | 2714 | 86 | ||
| L24434.1 | - | 21 - 22 | 2711 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thbs3
Genomic Map