Assay Details
Gene Symbol: Sgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291152.1 | NP_001278081.1 | 3 - 4 | 355 | 87 | |
NM_001291154.1 | NP_001278083.1 | 4 - 5 | 1071 | 87 | ||
NM_013731.3 | NP_038759.1 | 4 - 5 | 470 | 87 | ||
GenBank mRNA | AF169033.1 | - | 4 - 5 | 405 | 87 | |
AK050009.1 | - | 4 - 5 | 1052 | 87 | ||
AK136559.1 | - | 4 - 5 | 422 | 87 | ||
BC026549.1 | - | 2 - 3 | 100 | 4973908 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
