Assay Details
| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013791.2 | NP_038819.1 | 4 - 5 | 434 | 116 | |
| GenBank mRNA | AK037967.1 | - | 4 - 5 | 435 | 116 | |
| AK077541.1 | - | 4 - 5 | 430 | 116 | ||
| AK164822.1 | - | 4 - 5 | 430 | 116 | ||
| AK165239.1 | - | 4 - 5 | 434 | 116 | ||
| BC013703.1 | - | 4 - 5 | 418 | 3668925 | 116 | |
| U72194.1 | - | 4 - 5 | 417 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkln1
Genomic Map