Assay Details
| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013791.2 | NP_038819.1 | 7 - 8 | 811 | 71 | |
| GenBank mRNA | AK037967.1 | - | 7 - 8 | 812 | 71 | |
| BC013703.1 | - | 7 - 8 | 795 | 3668925 | 71 | |
| U72194.1 | - | 7 - 8 | 794 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkln1
Genomic Map