Assay Details
| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013791.2 | NP_038819.1 | 10 - 11 | 1195 | 79 | |
| GenBank mRNA | AK077541.1 | - | 10 - 11 | 1190 | 79 | |
| AK136123.1 | - | 7 - 8 | 880 | 79 | ||
| BC013703.1 | - | 10 - 11 | 1179 | 3668925 | 79 | |
| U72194.1 | - | 10 - 11 | 1178 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkln1
Genomic Map