Assay Details
| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013791.2 | NP_038819.1 | 15 - 16 | 1962 | 119 | |
| GenBank mRNA | AK077541.1 | - | 15 - 16 | 1957 | 119 | |
| BC013703.1 | - | 15 - 16 | 1946 | 3668925 | 119 | |
| CF161946.1 | - | 3 - 4 | 302 | 119 | ||
| U72194.1 | - | 15 - 16 | 1945 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkln1
Genomic Map