Assay Details
| Gene Symbol: Rnf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016698.2 | NP_057907.2 | 10 - 11 | 2128 | 69 | |
| GenBank mRNA | AB026621.1 | - | 10 - 11 | 2127 | 69 | |
| AK004579.1 | - | 10 - 11 | 2126 | 69 | ||
| AK034953.1 | - | 10 - 11 | 2127 | 69 | ||
| AK129106.1 | - | 10 - 11 | 2116 | 69 | ||
| AK146728.1 | - | 10 - 11 | 2128 | 69 | ||
| AK149972.1 | - | 10 - 11 | 2129 | 69 | ||
| AK156502.1 | - | 10 - 11 | 2128 | 69 | ||
| AK165617.1 | - | 10 - 11 | 2128 | 69 | ||
| AK168523.1 | - | 10 - 11 | 2123 | 69 | ||
| AK172065.1 | - | 10 - 11 | 2126 | 69 | ||
| BC010342.1 | - | 10 - 11 | 1837 | 3602969 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf10
Genomic Map