Assay Details
| Gene Symbol: Rnf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016698.2 | NP_057907.2 | 14 - 15 | 2601 | 80 | |
| GenBank mRNA | AB026621.1 | - | 14 - 15 | 2600 | 80 | |
| AK004579.1 | - | 14 - 15 | 2599 | 80 | ||
| AK034953.1 | - | 14 - 15 | 2600 | 80 | ||
| AK129106.1 | - | 14 - 15 | 2589 | 80 | ||
| AK146728.1 | - | 14 - 15 | 2601 | 80 | ||
| AK149972.1 | - | 14 - 15 | 2602 | 80 | ||
| AK156502.1 | - | 14 - 15 | 2601 | 80 | ||
| AK165617.1 | - | 14 - 15 | 2601 | 80 | ||
| AK168523.1 | - | 14 - 15 | 2596 | 80 | ||
| AK172065.1 | - | 14 - 15 | 2599 | 80 | ||
| AK187233.1 | - | 2 - 3 | 214 | 80 | ||
| AK218917.1 | - | 2 - 3 | 213 | 80 | ||
| BC010342.1 | - | 14 - 15 | 2310 | 3602969 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf10
Genomic Map