Assay Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016843.3 | NP_058539.2 | 2 - 3 | 393 | 115 | |
| GenBank mRNA | AK002504.1 | - | 2 - 3 | 502 | 115 | |
| AK003530.1 | - | 2 - 3 | 320 | 115 | ||
| AK047170.1 | - | 2 - 3 | 443 | 115 | ||
| AK049706.1 | - | 2 - 3 | 437 | 115 | ||
| AK077738.1 | - | 2 - 3 | 395 | 115 | ||
| AK078459.1 | - | 2 - 3 | 473 | 115 | ||
| AK153271.1 | - | 2 - 3 | 395 | 115 | ||
| AK161950.1 | - | 410 | 115 | |||
| AK165941.1 | - | 2 - 3 | 418 | 115 | ||
| AK166899.1 | - | 2 - 3 | 478 | 115 | ||
| AK170507.1 | - | 2 - 3 | 410 | 115 | ||
| AK170751.1 | - | 2 - 3 | 419 | 115 | ||
| BC016410.1 | - | 2 - 3 | 403 | 3968016 | 115 | |
| BC046802.1 | - | 2 - 3 | 529 | 6404607 | 115 | |
| X61506.1 | - | 2 - 3 | 507 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn10
Genomic Map