Assay Details
| Gene Symbol: Npff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018787.1 | NP_061257.1 | 2 - 3 | 256 | 81 | |
| NR_033409.1 | 1 - 2 | 812 | 81 | |||
| GenBank mRNA | AF148701.1 | - | 2 - 3 | 256 | 81 | |
| BC043025.1 | - | 1 - 2 | 812 | 81 | ||
| BC107294.1 | - | 1 - 2 | 486 | 40053067 | 81 | |
| BC107295.1 | - | 1 - 2 | 491 | 40053068 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
positive regulation of cytosolic calcium ion concentration
neuropeptide signaling pathway
negative regulation of heart rate
vasopressin secretion
negative regulation of appetite
positive regulation of blood pressure
negative regulation of insulin secretion
regulation of sensory perception of pain
excitatory postsynaptic potential
somatostatin secretion
G-protein coupled receptor binding
receptor binding
neuropeptide hormone activity
regulation of membrane depolarization
positive regulation of cytosolic calcium ion concentration
neuropeptide signaling pathway
negative regulation of heart rate
vasopressin secretion
negative regulation of appetite
positive regulation of blood pressure
negative regulation of insulin secretion
regulation of sensory perception of pain
excitatory postsynaptic potential
somatostatin secretion
G-protein coupled receptor binding
receptor binding
neuropeptide hormone activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | BC107294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | BC107295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | NM_018787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | NR_033409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | AF148701.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | BC107294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | BC107295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | NM_018787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | NR_033409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P232708 | AF148701.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148057 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146104 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | BC107294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | BC107295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | NM_018787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | NR_033409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | AF148701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | BC107294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | BC107295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | NM_018787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | NR_033409.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P232708 | AF148701.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | BC107294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | BC107295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | NM_018787.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | NR_033409.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | AF148701.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | BC107294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | BC107295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | NM_018787.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | NR_033409.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P232708 | AF148701.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30498431 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146104 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148057 | - | - |
| Whole Mouse Genome G4122A | A_51_P232708 | BC107294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | BC107295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | NM_018787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | NR_033409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | AF148701.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | BC107294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | BC107295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | NM_018787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | NR_033409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P232708 | AF148701.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109426 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146104 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148057 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11980965 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11980975 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12417396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12417387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11980967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11980969 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11980966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12027687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12331361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12110671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12110673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12027708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12032254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12027682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12027692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12331362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12417370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12032255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12027702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11980967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12319132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30498404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12027696 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10433030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433031 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433028 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420606_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433027 | - | - |
| Mouse Genome 430 2.0 Array | 1420606_at | - | - |
| Mouse Genome 430 2.0 Array | 1420606_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54615_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54615_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500018997.mm.1 | TC1500002295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018991.mm.1 | NM_018787.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018991.mm.1 | NM_018787.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018991.mm.1 | TC1500002295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018993.mm.1 | TC1500002295.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500018996.mm.1 | TC1500002295.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420606_at | - | - |
| Mouse Expression Set 430 | 1420606_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420606_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420606_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420606_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420606_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 853836 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2030280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2677157 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2677157 | - | - |
| mouse-6 v1.1 (Illumina) | scl054615.2_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030280 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl054615.2_19-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2677157 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2677157 | - | - |

Npff
Genomic Map