Assay Details
| Gene Symbol: Msln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018857.1 | NP_061345.1 | 9 - 10 | 1006 | 78 | |
| GenBank mRNA | AK144391.1 | - | 10 - 11 | 938 | 78 | |
| AK146799.1 | - | 9 - 10 | 1056 | 78 | ||
| BC023753.1 | - | 9 - 10 | 1010 | 5354484 | 78 | |
| D86370.1 | - | 9 - 10 | 1006 | 78 | ||
More Information
Set Membership: |
|

Msln
Genomic Map