Assay Details
| Gene Symbol: Mknk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285487.1 | NP_001272416.1 | 7 - 8 | 657 | 90 | |
| NM_001285488.1 | NP_001272417.1 | 6 - 7 | 551 | 90 | ||
| NM_021461.5 | NP_067436.1 | 5 - 6 | 507 | 90 | ||
| GenBank mRNA | AK010697.1 | - | 5 - 6 | 485 | 90 | |
| AK141298.1 | - | 5 - 6 | 485 | 90 | ||
| AK155934.1 | - | 7 - 8 | 618 | 90 | ||
| BC021369.1 | - | 5 - 6 | 463 | 5030044 | 90 | |
| BQ551007.1 | - | 3 - 4 | 214 | 90 | ||
| Y11091.1 | - | 5 - 6 | 472 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mknk1
Genomic Map