| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100011235.mm.1 |
NM_022018.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011235.mm.1 |
NM_022018.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011235.mm.1 |
TC0100001356.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011233.mm.1 |
NM_022018.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011233.mm.1 |
NM_022018.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011233.mm.1 |
TC0100001356.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011232.mm.1 |
NM_022018.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011232.mm.1 |
NM_022018.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011232.mm.1 |
TC0100001356.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011230.mm.1 |
NM_022018.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011230.mm.1 |
NM_022018.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011230.mm.1 |
TC0100001356.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011234.mm.1 |
NM_022018.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011234.mm.1 |
NM_022018.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011234.mm.1 |
TC0100001356.mm.1 |
MTA TC |