Assay Details
| Gene Symbol: Slc13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022411.3 | NP_071856.1 | 6 - 7 | 878 | 62 | |
| GenBank mRNA | AF201903.1 | - | 6 - 7 | 878 | 62 | |
| AK140972.1 | - | 6 - 7 | 928 | 62 | ||
| BC013493.1 | - | 6 - 7 | 903 | 4218337 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc13a2
Genomic Map