| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400012827.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012827.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012827.mm.1 |
TC1400001672.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400012819.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012819.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012819.mm.1 |
TC1400001672.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400012826.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012826.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012826.mm.1 |
TC1400001672.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400012828.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012828.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012828.mm.1 |
TC1400001672.mm.1 |
MTA TC |