| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400012791.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012791.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012791.mm.1 |
TC1400001672.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400012787.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012787.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012787.mm.1 |
TC1400001672.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400012789.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012789.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012789.mm.1 |
TC1400001672.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400012790.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012790.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012790.mm.1 |
TC1400001672.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400012793.mm.1 |
NM_022656.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012793.mm.1 |
NM_022656.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400012793.mm.1 |
TC1400001672.mm.1 |
MTA TC |