Assay Details
| Gene Symbol: Recql | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204906.1 | NP_001191835.1 | 8 - 9 | 1043 | 147 | |
| NM_001204907.1 | NP_001191836.1 | 8 - 9 | 1043 | 147 | ||
| NM_023042.3 | NP_075529.2 | 8 - 9 | 1043 | 147 | ||
| GenBank mRNA | AB017104.1 | - | 7 - 8 | 1004 | 147 | |
| AB017105.1 | - | 7 - 8 | 996 | 147 | ||
| AK137589.1 | - | 8 - 9 | 1043 | 147 | ||
| AK138325.1 | - | 8 - 9 | 999 | 147 | ||
| AK138721.1 | - | 8 - 9 | 1036 | 147 | ||
| AK148430.1 | - | 8 - 9 | 1031 | 147 | ||
| AK164344.1 | - | 8 - 9 | 1013 | 147 | ||
| BC014735.1 | - | 7 - 8 | 975 | 4459244 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Recql
Genomic Map