Assay Details
| Gene Symbol: Mrpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024177.3 | NP_077139.2 | 4 - 5 | 685 | 82 | |
| GenBank mRNA | AK088343.1 | - | 4 - 5 | 624 | 82 | |
| AK151598.1 | - | 4 - 5 | 624 | 82 | ||
| AK159033.1 | - | 4 - 5 | 686 | 82 | ||
| BC002319.1 | - | 3 - 4 | 661 | 3593984 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl38
Genomic Map