Assay Details
| Gene Symbol: Mrpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024177.3 | NP_077139.2 | 6 - 7 | 803 | 62 | |
| GenBank mRNA | AK088343.1 | - | 6 - 7 | 742 | 62 | |
| AK151598.1 | - | 6 - 7 | 742 | 62 | ||
| AK159033.1 | - | 6 - 7 | 804 | 62 | ||
| BC002319.1 | - | 5 - 6 | 779 | 3593984 | 62 | |
| BC030739.1 | - | 6 - 7 | 730 | 1493919 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl38
Genomic Map