Assay Details
Gene Symbol: Trim39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024468.2 | NP_077788.2 | 1 - 2 | 104 | 74 | |
GenBank mRNA | AB046382.1 | - | 1 - 2 | 104 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB046382.1,NM_024468.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
