Assay Details
| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144500.3 | NP_653083.2 | 8 - 9 | 978 | 90 | |
| GenBank mRNA | AK048537.1 | - | 8 - 9 | 898 | 90 | |
| AK145292.1 | - | 8 - 9 | 1010 | 90 | ||
| AK161695.1 | - | 8 - 9 | 917 | 90 | ||
| AK166059.1 | - | 8 - 9 | 957 | 90 | ||
| AK169504.1 | - | 1 - 2 | 903 | 90 | ||
| AK170306.1 | - | 8 - 9 | 962 | 90 | ||
| AK170669.1 | - | 8 - 9 | 979 | 90 | ||
| AK178638.1 | - | 2 - 3 | 157 | 90 | ||
| AK220257.1 | - | 2 - 3 | 297 | 90 | ||
| BC026804.1 | - | 8 - 9 | 985 | 4159217 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl2
Genomic Map