Assay Details
| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144500.3 | NP_653083.2 | 11 - 12 | 1330 | 73 | |
| GenBank mRNA | AK048537.1 | - | 11 - 12 | 1250 | 73 | |
| AK145292.1 | - | 11 - 12 | 1362 | 73 | ||
| AK166059.1 | - | 11 - 12 | 1309 | 73 | ||
| AK170306.1 | - | 11 - 12 | 1314 | 73 | ||
| AK170669.1 | - | 11 - 12 | 1331 | 73 | ||
| AK220257.1 | - | 5 - 6 | 649 | 73 | ||
| BC026804.1 | - | 11 - 12 | 1337 | 4159217 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl2
Genomic Map