Assay Details
| Gene Symbol: Sftpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282071.2 | NP_001269000.1 | 7 - 8 | 777 | 79 | |
| NM_147779.2 | NP_680088.1 | 7 - 8 | 846 | 79 | ||
| GenBank mRNA | AI430444.1 | - | 4 - 5 | 430 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P269134 | NM_147779.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P269134 | NM_147779.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096266 | NM_147779.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57836 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197508 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821151 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131043 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114816 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096266 | NM_147779.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P269134 | NM_147779.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P269134 | NM_147779.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269134 | NM_147779.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P269134 | NM_147779.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24540965 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114816 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01354200 | - | - |
| Whole Mouse Genome G4122A | A_51_P269134 | NM_147779.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P269134 | NM_147779.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124153 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24541018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24540940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05092436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05116014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05380265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05234123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05074729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05380264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05609877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05380255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05301878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05234118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05609868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05380260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05234130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05011558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05260997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05854087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05384446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05234125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05384450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24541003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24540952 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539094 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539092 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437028_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539091 | - | - |
| Mouse Genome 430 2.0 Array | 1437028_at | - | - |
| Mouse Genome 430 2.0 Array | 1437028_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20388_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20388_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600006452.mm.1 | NM_001282071.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006452.mm.1 | TC0600000846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006449.mm.1 | NM_001282071.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006449.mm.1 | TC0600000846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006451.mm.1 | NM_001282071.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006451.mm.1 | TC0600000846.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437028_PM_at | - | - |
| Mouse Expression Set 430 | 1437028_at | - | - |
| Murine Genome U74v2 Set | 160891_at | - | - |
| Murine Genome U74v2 Set | 160891_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437028_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437028_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437028_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437028_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3130603 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3130603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733762 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2733762 | - | - |
| mouse-6 v1.1 (Illumina) | scl29886.6_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130603 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29886.6_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2733762 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2792670 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733762 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2792670 | - | - |

Sftpb
Genomic Map