Assay Details
Gene Symbol: Usp9y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148943.2 | NP_683745.2 | 23 - 24 | 3572 | 129 | |
GenBank mRNA | AJ307017.1 | - | 23 - 24 | 3572 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
