Assay Details
| Gene Symbol: Cyp2b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009999.4 | NP_034129.1 | 4 - 5 | 671 | 88 | |
| GenBank mRNA | AF128849.1 | - | 4 - 5 | 653 | 88 | |
| AF129405.1 | - | 3 - 4 | 467 | 88 | ||
| AK028103.1 | - | 4 - 5 | 671 | 88 | ||
| BC060973.1 | - | 4 - 5 | 665 | 30301385 | 88 | |
| M21856.1 | - | 4 - 5 | 653 | 88 | ||
| X99715.1 | - | 4 - 5 | 649 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2b10
Genomic Map