Assay Details
| Gene Symbol: Rhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011270.3 | NP_035400.3 | 7 - 8 | 1137 | 72 | |
| GenBank mRNA | AB015189.1 | - | 7 - 8 | 1072 | 72 | |
| AF012430.1 | - | 7 - 8 | 1052 | 72 | ||
| AF047827.1 | - | 7 - 8 | 1072 | 72 | ||
| AF057524.1 | - | 7 - 8 | 1089 | 72 | ||
| AF069311.1 | - | 7 - 8 | 1091 | 72 | ||
| AK079335.1 | - | 7 - 8 | 1081 | 72 | ||
| AK157850.1 | - | 7 - 8 | 1064 | 72 | ||
| AK166030.1 | - | 5 - 6 | 820 | 72 | ||
| AK171641.1 | - | 7 - 8 | 1078 | 72 | ||
| BC145805.1 | - | 7 - 8 | 1168 | 40131108 | 72 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BG791443.1 | - | Chr. - - [-] on Build | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rhd
Genomic Map