Assay Details
| Gene Symbol: Rhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011270.3 | NP_035400.3 | 8 - 9 | 1222 | 64 | |
| GenBank mRNA | AB015189.1 | - | 8 - 9 | 1157 | 64 | |
| AF012430.1 | - | 8 - 9 | 1137 | 64 | ||
| AF047827.1 | - | 8 - 9 | 1157 | 64 | ||
| AF057524.1 | - | 8 - 9 | 1174 | 64 | ||
| AF069311.1 | - | 8 - 9 | 1176 | 64 | ||
| AK079335.1 | - | 8 - 9 | 1166 | 64 | ||
| AK157850.1 | - | 8 - 9 | 1149 | 64 | ||
| AK171641.1 | - | 8 - 9 | 1163 | 64 | ||
| BC145805.1 | - | 8 - 9 | 1253 | 40131108 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhd
Genomic Map