| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200007745.mm.1 |
NM_001029843.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007745.mm.1 |
NM_001029843.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007745.mm.1 |
NM_001029844.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007745.mm.1 |
NM_001029844.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007745.mm.1 |
NM_011705.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007745.mm.1 |
NM_011705.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007745.mm.1 |
TC1200001077.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200007744.mm.1 |
NM_001029843.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007744.mm.1 |
NM_001029843.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007744.mm.1 |
NM_001029844.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007744.mm.1 |
NM_001029844.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007744.mm.1 |
NM_011705.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007744.mm.1 |
NM_011705.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007744.mm.1 |
TC1200001077.mm.1 |
MTA TC |