Assay Details
| Gene Symbol: Pigq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291025.1 | NP_001277954.1 | 1 - 2 | 177 | 88 | |
| NM_011822.4 | NP_035952.2 | 1 - 2 | 177 | 88 | ||
| GenBank mRNA | AB008895.1 | - | 1 - 2 | 132 | 88 | |
| AK016852.1 | - | 1 - 2 | 124 | 88 | ||
| AK033349.1 | - | 1 - 2 | 124 | 88 | ||
| AK137390.1 | - | 1 - 2 | 118 | 88 | ||
| AK155173.1 | - | 1 - 2 | 80 | 88 | ||
| AK163680.1 | - | 1 - 2 | 153 | 88 | ||
| AK169284.1 | - | 1 - 2 | 160 | 88 | ||
| AK170431.1 | - | 1 - 2 | 124 | 88 | ||
| AK193861.1 | - | 1 - 2 | 69 | 88 | ||
| BC014287.1 | - | 1 - 2 | 150 | 4193343 | 88 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB008895.1,AK163680.1,AK169284.1,AK170431.1,BC014287.1,NM_001291025.1,NM_011822.4 transcripts.
Set Membership: |
|

Pigq
Genomic Map