Assay Details
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291025.1 | NP_001277954.1 | 3 - 4 | 1007 | 61 | |
NM_011822.4 | NP_035952.2 | 3 - 4 | 1007 | 61 | ||
GenBank mRNA | AB008895.1 | - | 3 - 4 | 962 | 61 | |
AK016852.1 | - | 3 - 4 | 954 | 61 | ||
AK033349.1 | - | 3 - 4 | 954 | 61 | ||
AK155173.1 | - | 3 - 4 | 910 | 61 | ||
AK163680.1 | - | 3 - 4 | 983 | 61 | ||
AK169284.1 | - | 3 - 4 | 990 | 61 | ||
AK170431.1 | - | 3 - 4 | 954 | 61 | ||
AK218878.1 | - | 2 - 3 | 200 | 61 | ||
BC003917.1 | - | 1 - 2 | 97 | 61 | ||
BC014287.1 | - | 3 - 4 | 980 | 4193343 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
