Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400003024.mm.1 |
NM_013497.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003024.mm.1 |
NM_013497.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003024.mm.1 |
TC0400000439.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003020.mm.1 |
NM_013497.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003020.mm.1 |
NM_013497.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003020.mm.1 |
TC0400000439.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003019.mm.1 |
NM_013497.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003019.mm.1 |
NM_013497.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003019.mm.1 |
TC0400000439.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400003021.mm.1 |
NM_013497.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003021.mm.1 |
NM_013497.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400003021.mm.1 |
TC0400000439.mm.1 |
MTA TC |