Assay Details
| Gene Symbol: St8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013666.2 | NP_038694.2 | 2 - 3 | 485 | 88 | |
| GenBank mRNA | BY733136.1 | - | 2 - 3 | 467 | 88 | |
| X98014.1 | - | 2 - 3 | 348 | 88 | ||
More Information
Set Membership: |
|

St8sia5
Genomic Map