Assay Details
| Gene Symbol: Dclre1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018831.4 | NP_061301.3 | 3 - 4 | 2737 | 121 | |
| GenBank mRNA | AF241240.1 | - | 2 - 3 | 2202 | 121 | |
| AK029794.1 | - | 2 - 3 | 2406 | 121 | ||
| AK220214.1 | - | 2 - 3 | 1959 | 121 | ||
| BC031858.1 | - | 1 - 2 | 1628 | 121 | ||
| BC072667.1 | - | 2 - 3 | 2153 | 30629417 | 121 | |
| BC076617.1 | - | 3 - 4 | 2737 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dclre1a
Genomic Map