Assay Details
| Gene Symbol: Dclre1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018831.4 | NP_061301.3 | 4 - 5 | 2870 | 85 | |
| GenBank mRNA | AF241240.1 | - | 3 - 4 | 2335 | 85 | |
| AK029794.1 | - | 3 - 4 | 2539 | 85 | ||
| AK220214.1 | - | 3 - 4 | 2092 | 85 | ||
| BC031858.1 | - | 2 - 3 | 1761 | 85 | ||
| BC072667.1 | - | 3 - 4 | 2286 | 30629417 | 85 | |
| BC076617.1 | - | 4 - 5 | 2870 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dclre1a
Genomic Map